Läs mer
2-Methoxytetrahydrofuran, TRC
Produktkod. 30481215
Change view
Kvantitet:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30481215 1 g
30466755 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30481215 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM2645051G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 13436-45-8
Molekylformel C5H10O2
Molekylvikt (g/mol) 102.13
Synonym Tetrahydro-2-methoxyfuran
IUPAC-namn 2-methoxyoxolane
LEDER COC1CCCO1
Specifikationer
Kemiskt namn eller material 2-Methoxytetrahydrofuran
Kvantitet 1 g
InChI formel InChI=1S/C5H10O2/c1-6-5-3-2-4-7-5/h5H,2-4H2,1H3
Formel vikt 102.0681