missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1,3-cyclopentanedione, 98%, Thermo Scientific Chemicals

Catalog Number p-161745
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
765-69-5
C6H8O2
112.13
MFCD00001406
HXZILEQYFQYQCE-UHFFFAOYSA-N
2-methyl-1,3-cyclopentanedione, 1,3-cyclopentanedione, 2-methyl, 2-methyl-1,3-cyclopentadione, 2-methyl-cyclopentane-1,3-dione, unii-c9lg5vp01c, c9lg5vp01c, methylcycle-d, pubchem2206, acmc-209p4n, benzil-related compound, 46
13005
2-methylcyclopentane-1,3-dione
CC1C(=O)CCC1=O
This item is not returnable. View return policy
765-69-5
C6H8O2
112.13
MFCD00001406
HXZILEQYFQYQCE-UHFFFAOYSA-N
2-methyl-1,3-cyclopentanedione, 1,3-cyclopentanedione, 2-methyl, 2-methyl-1,3-cyclopentadione, 2-methyl-cyclopentane-1,3-dione, unii-c9lg5vp01c, c9lg5vp01c, methylcycle-d, pubchem2206, acmc-209p4n, benzil-related compound, 46
13005
2-methylcyclopentane-1,3-dione
CC1C(=O)CCC1=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 212.0°C to 216.0°C
Color Beige to Tan
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 25 g
Solubility Information Solubility in water: .. Other solubilities: soluble in methanol
Formula Weight 112.13
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-Methyl-1, 3-cyclopentanedione, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'