missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1,3-thiazole-4-carbothioamide, 97%, Thermo Scientific™

Catalog Number p-2698566
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
174223-29-1
C5H6N2S2
158.237
MFCD00052167
DIOPPULTIGEDCB-UHFFFAOYSA-N
2-methylthiazole-4-carbothioamide, amino 2-methyl 1,3-thiazol-4-yl methane-1-thione, maybridge4_004334, 2-methylthiazole-4-thiocarboxamide, 4-thiazolecarbothioamide,2-methyl, 4-thiazolecarbothioamide, 2-methyl, 2-methylthiazole-4-carbothioic acid amide, 4-thiazolecarbothioamide, 2-methyl-9ci
2744365
2-methyl-1,3-thiazole-4-carbothioamide
CC1=NC(=CS1)C(=S)N
This item is not returnable. View return policy
174223-29-1
C5H6N2S2
158.237
MFCD00052167
DIOPPULTIGEDCB-UHFFFAOYSA-N
2-methylthiazole-4-carbothioamide, amino 2-methyl 1,3-thiazol-4-yl methane-1-thione, maybridge4_004334, 2-methylthiazole-4-thiocarboxamide, 4-thiazolecarbothioamide,2-methyl, 4-thiazolecarbothioamide, 2-methyl, 2-methylthiazole-4-carbothioic acid amide, 4-thiazolecarbothioamide, 2-methyl-9ci
2744365
2-methyl-1,3-thiazole-4-carbothioamide
CC1=NC(=CS1)C(=S)N
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 158.25
Percent Purity 97%
Chemical Name or Material 2-Methyl-1, 3-thiazole-4-carbothioamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'