Läs mer
2-Methyl-1h,2h,3h,4h,9h-pyrido[3,4-b]indole, TRC
Produktkod. 30003250
Change view
Kvantitet:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30003250 25 mg
30007019 10 mg
30004997 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30003250 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM20336125MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 13100-00-0
Molekylformel C12 H14 N2
Molekylvikt (g/mol) 186.253
Synonym 2,3,4,9-Tetrahydro-2-methyl-1H-pyrido[3,4-b]indole, 1,2,3,4-Tetrahydro-2-methyl-9H-pyrido[3,4-b]indole, 2-Methyl-1,2,3,4-tetrahydro-β-carboline, 2-Methyl-1,3,4,9-tetrahydropyrido[3,4-b]indole, 2-Methyltetrahydro-β-carboline, N-Methyl-1,2,3,4-tetrahydro-β-carboline, NSC 683435;
IUPAC-namn 2-methyl-1,3,4,9-tetrahydropyrido[3,4-b]indole
LEDER CN1CC2=C(CC1)C3C(=CC=CC=3)N2
Specifikationer
Kemiskt namn eller material 2-Methyl-1h,2h,3h,4h,9h-pyrido[3,4-b]indole
Kvantitet 25 mg
InChI formel InChI=1S/C12H14N2/c1-14-7-6-10-9-4-2-3-5-11(9)13-12(10)8-14/h2-5,13H,6-8H2,1H3
Formel vikt 186.116