Learn More
2-Methyl-1H-benzimidazole-5-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals
Catalog Number p-4602058
Click to view available options
Quantity:
1 g
5 g
1314216-34-6
C14H19BN2O2
258.13
MFCD13176536
CPQGLMCLTALRHF-UHFFFAOYSA-N
2-methyl-1h-benzimidazole-5-boronic acid pinacol ester, 2-methyl-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-benzo d imidazole, 2-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-benzo d imidazole, cpqglmcltalrhf-uhfffaoysa-n, 2-methyl-1h-benzimidazole-5-boronic acidpinacol ester, 2-methyl-1h-benzimidazole-5-boronic acid, pinacol ester, 1h-benzimidazole, 2-methyl-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 2-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-benzo d imidazole, 2-methyl-6-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1h-benzo d imidazole, 2-methyl-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-benzimidazole
46738035
2-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-1,3-benzodiazole
CC1=NC2=CC(=CC=C2N1)B1OC(C)(C)C(C)(C)O1
1314216-34-6
C14H19BN2O2
258.13
MFCD13176536
CPQGLMCLTALRHF-UHFFFAOYSA-N
2-methyl-1h-benzimidazole-5-boronic acid pinacol ester, 2-methyl-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-benzo d imidazole, 2-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-benzo d imidazole, cpqglmcltalrhf-uhfffaoysa-n, 2-methyl-1h-benzimidazole-5-boronic acidpinacol ester, 2-methyl-1h-benzimidazole-5-boronic acid, pinacol ester, 1h-benzimidazole, 2-methyl-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 2-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-benzo d imidazole, 2-methyl-6-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1h-benzo d imidazole, 2-methyl-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-benzimidazole
46738035
2-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-1,3-benzodiazole
CC1=NC2=CC(=CC=C2N1)B1OC(C)(C)C(C)(C)O1
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 211°C to 212°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Quantity 1 g
Formula Weight 258.13
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-Methyl-1H-benzimidazole-5-boronic acid pinacol ester
compliance-icons
Product Identifier
  • 2-Methyl-1H-benzimidazole-5-boronic acid pinacol ester
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.