Läs mer
2-Methyl-1H-quinolin-4-one, TRC
Produktkod. 30008859
Change view
Kvantitet:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30008859 250 mg
30008692 100 mg
30006170 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30008859 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM396080250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 5660-24-2
Molekylformel C10H9NO
Molekylvikt (g/mol) 159.18
Synonym 2-Methyl-4(1H)-quinolinone, 2-Methyl-1,4-dihydro-4-oxoquinoline, 2-Methyl-1,4-dihydroquinolin-4-one, 2-Methyl-1H-quinolin-4-one, 2-Methyl-4(1H)-quinolone, 2-Methyl-4-quinolinone, 4(1H)-Quinaldone
IUPAC-namn 2-methyl-1H-quinolin-4-one
LEDER CC1=CC(=O)c2ccccc2N1
Specifikationer
Kemiskt namn eller material 2-Methyl-1H-quinolin-4-one
Kvantitet 250 mg
InChI formel InChI=1S/C10H9NO/c1-7-6-10(12)8-4-2-3-5-9(8)11-7/h2-6H,1H3,(H,11,12)
Formel vikt 159.0684