missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyl-2-phenylpropylmagnesium chloride, 0.5M solution in diethyl ether, AcroSeal™, Thermo Scientific Chemicals
599.60 SEK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
CAS Min % | 10.16, 84.77 |
CAS Max % | 15.23, 89.84 |
Color | Colorless to Yellow |
Density | 0.7600g/mL |
Flash Point | −28°C |
Packaging | AcroSeal™ Glass Bottle |
Linear Formula | C6H5C(CH3)2CH2MgCl |
Quantity | 100 mL |
Specific Gravity | 0.76 |
Formula Weight | 192.97 |
Show More |
Safety and Handling
Product Identifier
- 2-Methyl-2-phenylpropylmagnesium chloride, 0.5M solution in diethyl ether
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H336-May cause drowsiness or dizziness.
- H302-Harmful if swallowed.
- H224-Extremely flammable liquid and vapour.
- EUH014-Reacts violently with water.
- EUH019-May form explosive peroxides.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'