missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-2H-1,4-benzoxazin-3(4H)-one, 97%, Thermo Scientific™

Catalog Number p-7058140
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
21744-83-2
C9H9NO2
163.176
MFCD04141926
PILVMCZSXIINLV-UHFFFAOYSA-N
2-methyl-2h-benzo b 1,4 oxazin-3 4h-one, 2-methyl-2h-1,4-benzoxazin-3 4h-one, 2-methyl-1,4-benzoxazin-3-one, 2-methyl-1,4-benzoxazin-3-4h-one, 2-methyl-3,4-dihydro-2h-1,4-benzoxazin-3-one, 2-methyl-2,4-dihydro-1,4-benzoxazin-3-one, acmc-1cjvq, 2-methyl-1,4 benzoxazin-3 4h-one, 2-methyl-4h-benzo 1,4 oxazin-3-one, 2h-1,4-benzoxazin-3 4h-one,2-methyl
301730
2-methyl-4H-1,4-benzoxazin-3-one
CC1C(=O)NC2=CC=CC=C2O1
This item is not returnable. View return policy
21744-83-2
C9H9NO2
163.176
MFCD04141926
PILVMCZSXIINLV-UHFFFAOYSA-N
2-methyl-2h-benzo b 1,4 oxazin-3 4h-one, 2-methyl-2h-1,4-benzoxazin-3 4h-one, 2-methyl-1,4-benzoxazin-3-one, 2-methyl-1,4-benzoxazin-3-4h-one, 2-methyl-3,4-dihydro-2h-1,4-benzoxazin-3-one, 2-methyl-2,4-dihydro-1,4-benzoxazin-3-one, acmc-1cjvq, 2-methyl-1,4 benzoxazin-3 4h-one, 2-methyl-4h-benzo 1,4 oxazin-3-one, 2h-1,4-benzoxazin-3 4h-one,2-methyl
301730
2-methyl-4H-1,4-benzoxazin-3-one
CC1C(=O)NC2=CC=CC=C2O1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 163.18
Percent Purity 97%
Chemical Name or Material 2-Methyl-2H-1,4-benzoxazin-3(4H)-one
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'