missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyl-3-nitroanisole, 99%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
Specifications
CAS Min % | 98.5 |
CAS Max % | 100.0 |
Melting Point | 54.0°C to 55.0°C |
Color | Brown to Yellow |
Flash Point | >112°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 98.5% min. (GC) |
Packaging | Glass Bottle |
Linear Formula | CH3C6H3(NO2)OCH3 |
Quantity | 1 g |
Show More |
Safety and Handling
Product Identifier
- 2-Methyl-3-nitroanisole
- Warning
- Acute toxicity Category 4
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'