Läs mer
2-Methyl-3-oxopentanoic Acid Methyl Ester, TRC
Produktkod. 30467804
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30467804 100 mg
1 options
Produktkod. 30467804 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM321075100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-Methyl-3-oxopentanoic Acid Methyl Ester
CAS 17422-12-7
Molekylformel C7H12O3
Kvantitet 100 mg
Synonym Methyl 2,4-Dimethyl-3-oxobutyrate, Methyl 2-Methyl-3-oxopentanoate, Methyl 2-Methyl-3-oxovalerate, Methyl 2-Propionylpropionate, Methyl 3-Oxo-2-methylpentanoate
InChI formel InChI=1S/C7H12O3/c1-4-6(8)5(2)7(9)10-3/h5H,4H2,1-3H3
LEDER O=C(C(C)C(CC)=O)OC
IUPAC-namn methyl 2-methyl-3-oxopentanoate
Molekylvikt (g/mol) 144.17
Formel vikt 144.08