Learn More
2-Methyl-3-(trifluoromethyl)aniline, 97%, BLD Pharm™
Product Code. 31004689
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
100 g
500 g
1 kg
Unit Size:
100g
10g
1g
1kg
25g
500g
5g
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31004689 25 g 25g
31004685 1 g 1g
30990224 100 g 100g
30998035 1 kg 1kg
30995972 10 g 10g
30995881 500 g 500g
30995535 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 31004689 Supplier BLD Pharm Supplier No. BD1112425g
 Restricted Item
Beräknad leverans: 27-11-2026
Specifications
CAS 54396-44-0
Molecular Formula C8H8F3N
MDL Number MFCD00153216
Quantity 25 g
SMILES NC1=CC=CC(C(F)(F)F)=C1C
Molecular Weight (g/mol) 175.15
Percent Purity 97%
Chemical Name or Material 2-Methyl-3-(trifluoromethyl)aniline