missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-3-(trifluoromethyl)benzoic acid, 97%, Thermo Scientific™

Catalog Number p-7029086
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
62089-35-4
C9H7F3O2
204.15
MFCD01631588
SSCFPSMXIDAJDT-UHFFFAOYSA-N
2-methyl-3-trifluoromethyl benzoic acid, benzoic acid, 2-methyl-3-trifluoromethyl, 2-methyl-3-trifluoromethyl benzoicacid, acmc-209ws1, ksc614g1p, 3-carboxy-2-methylbenzotrifluoride, 2-carboxy-6-trifluoromethyl toluene, 3-trifluoromethyl-2-methybenzoic acid, 3-trifluoromethyl-2-methylbenzoic acid, 2-methyl-3-trifluoromethyl-benzoic acid
2775584
2-methyl-3-(trifluoromethyl)benzoic acid
CC1=C(C=CC=C1C(F)(F)F)C(O)=O
This item is not returnable. View return policy
62089-35-4
C9H7F3O2
204.15
MFCD01631588
SSCFPSMXIDAJDT-UHFFFAOYSA-N
2-methyl-3-trifluoromethyl benzoic acid, benzoic acid, 2-methyl-3-trifluoromethyl, 2-methyl-3-trifluoromethyl benzoicacid, acmc-209ws1, ksc614g1p, 3-carboxy-2-methylbenzotrifluoride, 2-carboxy-6-trifluoromethyl toluene, 3-trifluoromethyl-2-methybenzoic acid, 3-trifluoromethyl-2-methylbenzoic acid, 2-methyl-3-trifluoromethyl-benzoic acid
2775584
2-methyl-3-(trifluoromethyl)benzoic acid
CC1=C(C=CC=C1C(F)(F)F)C(O)=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 106°C to 109°C
Quantity 1 g
Formula Weight 204.15
Percent Purity 97%
Chemical Name or Material 2-Methyl-3-(trifluoromethyl)benzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'