Läs mer
2-Methyl-3-tritylamino-4-[(2-N-bocamino)ethylaminoacarbonyl]amino-2H-pyrazole, TRC
Produktkod. 30468611
Change view
Kvantitet:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30468611 50 mg
30464870 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30468611 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM79030050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-Methyl-3-tritylamino-4-[(2-N-bocamino)ethylaminoacarbonyl]amino-2H-pyrazole
CAS 689293-69-4
Molekylformel C31 H36 N6 O3
Kvantitet 50 mg
Synonym Carbamic acid','[2-[[[[1-methyl-5-[(triphenylmethyl)amino]-1H-pyrazol-4-yl]amino]carbonyl]amino]ethyl]-','1,1-dimethylethyl ester (9CI), 1,1-Dimethylethyl N-[2-[[[[1-methyl-5-[(triphenylmethyl)amino]-1H-pyrazol-4-yl]amino]carbonyl]amino]ethyl]carbamate
InChI formel InChI=1S/C31H36N6O3/c1-30(2,3)40-29(39)33-21-20-32-28(38)35-26-22-34-37(4)27(26)36-31(23-14-8-5-9-15-23,24-16-10-6-11-17-24)25-18-12-7-13-19-25/h5-19,22,36H,20-21H2,1-4H3,(H,33,39)(H2,32,35,38)
LEDER Cn1ncc(NC(=O)NCCNC(=O)OC(C)(C)C)c1NC(c2ccccc2)(c3ccccc3)c4ccccc4
IUPAC-namn tert-butyl N-[2-[[1-methyl-5-(tritylamino)pyrazol-4-yl]carbamoylamino]ethyl]carbamate
Molekylvikt (g/mol) 540.656
Formel vikt 540.285