Läs mer
2-Methyl-4,6-bis(octylsulfanylmethyl)phenol, TRC
Produktkod. 30483983
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30483983 5 mg
1 options
Produktkod. 30483983 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM3257335MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2,4-Bis(octylthiomethyl)-6-methylphenol
CAS 110553-27-0
Molekylformel C25H44OS2
Kvantitet 5 mg
Synonym 2-Methyl-4,6-bis[(octylthio)methyl]phenol, 2,4-Bis(octylthiomethyl)-6-methylphenol, 2,4-Dioctylthiomethyl-6-methylphenol, 2-Methyl-4,6-Bis[(octylthio)methyl]phenol, 2-Methyl-4,6-bis(octylsulfanylmethyl)phenol, 4,6-Bis(octylthiomethyl)-o-cresol, 4,6-Bis(octythiomethyl)-o-cresol, AO 4, AO 4 (Russian stabilizer), Antage HP 400, Antioxidant 1520, Antioxidant 2088, EB 51-677, HP 400, IR 1520, IX 1520, Irganox 1520, Irganox 1520D, Irganox 1520L, Irganox 1520SE, Irgastab Cable KV 10, Irgastab KV 10, KV 10, S 6P
InChI formel InChI=1S/C25H44OS2/c1-4-6-8-10-12-14-16-27-20-23-18-22(3)25(26)24(19-23)21-28-17-15-13-11-9-7-5-2/h18-19,26H,4-17,20-21H2,1-3H3
LEDER CCCCCCCCSCc1cc(C)c(O)c(CSCCCCCCCC)c1
IUPAC-namn 2-methyl-4,6-bis(octylsulfanylmethyl)phenol
Molekylvikt (g/mol) 424.75
Formel vikt 424.2834