Läs mer
2-Methyl-4-isothiazolin-3-one Hydrochloride, TRC
Produktkod. 30040503
Change view
Kvantitet:
25 mg
1 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30040503 25 mg
30009359 1 g
30004162 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30040503 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM31498525MG
Denna vara kräver en prisuppdatering innan den beställs.
Begär ett pris för denna vara här.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 26172-54-3
Molekylformel C4 H5 N O S . Cl H
Molekylvikt (g/mol) 151.61
Synonym 2-Methyl-4-isothiazolin-3-one Hydrochloride, 2-Methylisothiazol-3-one hydrochloride, 3(2H)-Isothiazolone, 2-methyl-, hydrochloride (1:1), 3(2H)-Isothiazolone, 2-methyl-, hydrochloride (9CI), 4-Isothiazolin-3-one, 2-methyl-, hydrochloride (8CI), 2-Methyl-3-isothiazolone hydrochloride, 2-Methyl-4-isothiazolin-3-one hydrochloride
IUPAC-namn 2-methyl-1,2-thiazol-3-one;hydrochloride
LEDER Cl.CN1SC=CC1=O
Specifikationer
Kemiskt namn eller material 2-Methyl-4-isothiazolin-3-on hydrochloride
Kvantitet 25 mg
InChI formel InChI=1S/C4H5NOS.ClH/c1-5-4(6)2-3-7-5;/h2-3H,1H3;1H
Formel vikt 150.9859