Läs mer
2-Methyl-4-nitrophenylguanidine Nitrate, TRC
Produktkod. 30476940
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30476940 50 mg
1 options
Produktkod. 30476940 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM32540050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-Methyl-4-nitrophenylguanidine nitrate
CAS 796738-73-3
Molekylformel C8 H10 N4 O2 . H N O3
Kvantitet 50 mg
Synonym Guanidine, N-(2-methyl-4-nitrophenyl)-, nitrate (1:1), Guanidine, (2-methyl-4-nitrophenyl)-, mononitrate (9CI), 2-Methyl-4-nitrophenylguanidine nitrate
InChI formel InChI=1S/C8H10N4O2.HNO3/c1-5-4-6(12(13)14)2-3-7(5)11-8(9)10;2-1(3)4/h2-4H,1H3,(H4,9,10,11);(H,2,3,4)
LEDER Cc1cc(ccc1NC(=N)N)[N+](=O)[O-].O[N+](=O)[O-]
IUPAC-namn 1-(2-methyl-4-nitrophenyl)guanidine;nitric acid
Molekylvikt (g/mol) 257.2
Formel vikt 257.076