Läs mer
2-Methyl-5-nitro-1-(2-oxiranylmethyl)-1H-imidazole, TRC
Produktkod. 30133453
Change view
Kvantitet:
100 mg
1 g
2.5 g
Förpackningsstorlek:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30133453 2.5 g 2.5g
30133176 1 g -
30009569 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30133453 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM3201452.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 16773-52-7
Molekylformel C7 H9 N3 O3
Molekylvikt (g/mol) 183.16
Synonym 3-(2-Methyl-5-nitro-1H-imidazol-1-yl)-1,2-epoxypropane
IUPAC-namn 2-methyl-5-nitro-1-(oxiran-2-ylmethyl)imidazole
LEDER Cc1ncc([N+](=O)[O-])n1CC2CO2
Specifikationer
Kemiskt namn eller material 3-(2-Methyl-5-nitro-1H-imidazol-1-yl)-1,2-epoxypropane
Anteckningar om renhetsgrad HPLC
Kvantitet 2.5 g
InChI formel InChI=1S/C7H9N3O3/c1-5-8-2-7(10(11)12)9(5)3-6-4-13-6/h2,6H,3-4H2,1H3
Formel vikt 183.0644
Procent renhet >95%