missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyl-5-nitrobenzotrifluoride, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
Employed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Employed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field
Solubility
Slightly soluble in water.
Notes
Moisture Sensitive. Store away form water/moisture. Store away from oxidizing agent.
Employed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field
Solubility
Slightly soluble in water.
Notes
Moisture Sensitive. Store away form water/moisture. Store away from oxidizing agent.
Specifications
Color | White |
Quantity | 1 g |
UN Number | UN2811 |
Beilstein | 2506013 |
Sensitivity | Moisture sensitive |
Solubility Information | Slightly soluble in water. |
Formula Weight | 205.14 |
Percent Purity | 98% |
Physical Form | Solid |
Chemical Name or Material | 2-Methyl-5-nitrobenzotrifluoride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'