missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Methyl-5-nitropyridine, 95%

Brand:  Alfa Aesar™ H61750.14

Additional Details : CAS Number : 21203-68-9 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 15438318

  • 2724.50 SEK / 25g


Estimated Shipment
29-10-2020
Add to basket

Description

Description

Applications
2-Methyl-5-nitropyridine is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

21203-68-9
138.126
USZINSZJSVMICC-UHFFFAOYSA-N
2794552
CC1=NC=C(C=C1)[N+](=O)[O-]
C6H6N2O2
MFCD04114179
5-nitro-2-picoline, pyridine, 2-methyl-5-nitro, 5-nitro-2-methylpyridine, 2-methyl-5-nitro-pyridine, pubchem6699, acmc-1cewl, 2-methyl-5-nitro pyridine, ksc494i9p, abbypharma ap-15-5036
2-methyl-5-nitropyridine

Specifications

2-Methyl-5-nitropyridine
21203-68-9
MFCD04114179
Slightly soluble in water.
CC1=NC=C(C=C1)[N+](=O)[O-]
138.126
138.12
95%
25g
C6H6N2O2
5-nitro-2-picoline, pyridine, 2-methyl-5-nitro, 5-nitro-2-methylpyridine, 2-methyl-5-nitro-pyridine, pubchem6699, acmc-1cewl, 2-methyl-5-nitro pyridine, ksc494i9p, abbypharma ap-15-5036
USZINSZJSVMICC-UHFFFAOYSA-N
2-methyl-5-nitropyridine
2794552
Solid
Videos
SDS Documents