Läs mer
2-Methyl-5-(trifluoromethyl)benzaldehyde, 97%, Thermo Scientific™
Produktkod. 11322509
Change view
Kvantitet:
1 g
5 g
Förpackningsstorlek:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet Förpackningsstorlek
11322509 1 g 1g
11332509 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 11322509 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer H26628.03
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 886498-85-7
Molekylformel C9H7F3O
Molekylvikt (g/mol) 188.149
MDL-nummer MFCD06660200
InChI-nyckel XFIZZVVBLXLTOD-UHFFFAOYSA-N
Synonym 2-methyl-5-trifluoromethyl benzaldehyde, ksc496m3p
PubChem CID 17750779
IUPAC-namn 2-methyl-5-(trifluoromethyl)benzaldehyde
LEDER CC1=C(C=C(C=C1)C(F)(F)F)C=O
Specifikationer
Kemiskt namn eller material 2-Methyl-5-(trifluoromethyl)benzaldehyde
Brytningsindex 1.475
Kvantitet 1 g
Känslighet Air Sensitive
Formel vikt 188.15
Procent renhet 97%