missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyl-6-nitrobenzoic acid, 99%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
25 g
Specifications
Melting Point | 154.0°C to 157.0°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (HPLC) |
Packaging | Glass bottle |
Linear Formula | CH3C6H3(NO2)CO2H |
Quantity | 1 g |
Beilstein | 09,I,189 |
Solubility Information | Solubility in water: <0.1g/100mL (20°C). |
Formula Weight | 181.15 |
Percent Purity | 99% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'