Läs mer
2-Methyl-6-nitrobenzoic Acid Methyl Ester, TRC
Produktkod. 30467722
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30467722 1 g
1 options
Produktkod. 30467722 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM3202001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-Methyl-6-nitrobenzoic Acid Methyl Ester
CAS 61940-22-5
Molekylformel C9H9NO4
Kvantitet 1 g
Synonym Methyl 2-methyl-6-nitrobenzoate, Methyl 6-methyl-2-nitrobenzoate, 6-Nitro-o-toluic Acid Methyl Ester
InChI formel InChI=1S/C9H9NO4/c1-6-4-3-5-7(10(12)13)8(6)9(11)14-2/h3-5H,1-2H3
LEDER COC(=O)c1c(C)cccc1[N+](=O)[O-]
IUPAC-namn methyl 2-methyl-6-nitrobenzoate
Molekylvikt (g/mol) 195.17
Formel vikt 195.0532