Learn More
2-Methyl-6-nitrobenzoxazole, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
2-Methyl-6-nitrobenzoxazole is used as a flavoring agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Methyl-6-nitrobenzoxazole is used as a flavoring agents.
Solubility
Soluble in oils. Insoluble in water.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
2-Methyl-6-nitrobenzoxazole is used as a flavoring agents.
Solubility
Soluble in oils. Insoluble in water.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Melting Point | 154°C to 156°C |
Quantity | 1 g |
Synonym | 2-methyl-6-nitrobenzoxazole, benzoxazole, 2-methyl-6-nitro, acmc-1azgy, 2-methyl-6-nitro-benzoxazole, benzoxazole,2-methyl-6-nitro, 2-methyl-6-nitrobenzo d oxazole |
Solubility Information | Soluble in oils. Insoluble in water. |
Formula Weight | 146.17 |
Percent Purity | 98% |
Chemical Name or Material | 2-Methyl-6-nitrobenzoxazole |