Läs mer
2-Methyl Hippuric Acid, TRC
Produktkod. 30468978
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30468978 500 mg
1 options
Produktkod. 30468978 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM311980500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-Methyl Hippuric Acid
CAS 42013-20-7
Anteckningar om renhetsgrad HPLC
Molekylformel C10 H11 N O3
Kvantitet 500 mg
Synonym Glycine','N-(2-methylbenzoyl)-, N-(2-Methylbenzoyl)glycine, (2-Methyl-benzoylamino)-acetic acid, 2-Methylhippuric acid, 2-[(2-Methylbenzoyl)amino]acetic acid, 2-[(2-Methylphenyl)formamido]acetic acid, NSC 163983, o-Methylhippuric acid, o-Toluric acid, 2-Methyl hippuric acid
InChI formel InChI=1S/C10H11NO3/c1-7-4-2-3-5-8(7)10(14)11-6-9(12)13/h2-5H,6H2,1H3,(H,11,14)(H,12,13)
LEDER Cc1ccccc1C(=O)NCC(=O)O
IUPAC-namn 2-[(2-methylbenzoyl)amino]acetic acid
Molekylvikt (g/mol) 193.2
Formel vikt 193.0739