Läs mer
(-)-2-Methyl Isoborneol, TRC
Produktkod. 30136120
Change view
Kvantitet:
50 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136120 500 mg
30134882 1 g
30006055 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136120 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM314000500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 68330-43-8
Molekylformel C11H20O
Molekylvikt (g/mol) 168.28
Synonym (1R,2R,4R)-1,2,7,7-Tetramethylbicyclo[2.2.1]heptan-2-ol, (1R-exo)-1,2,7,7-Tetramethylbicyclo[2.2.1]heptan-2-ol, (-)-2-exo-Hydroxy-2-methylbornane, 2-methylisoborneol;
IUPAC-namn (1R,2R,4R)-1,2,7,7-tetramethylbicyclo[2.2.1]heptan-2-ol
LEDER O[C@@]1(C)[C@@]2(C)CC[C@H](C2(C)C)C1
Specifikationer
Kemiskt namn eller material (-)-2-Methyl Isoborneol
Kvantitet 500 mg
InChI formel InChI=1S/C11H20O/c1-9(2)8-5-6-10(9,3)11(4,12)7-8/h8,12H,5-7H2,1-4H3/t8-,10-,11-/m1/s1
Formel vikt 168.1514