Läs mer
2-methyl-N-phenylalanine, TRC
Produktkod. 30179496
Change view
Kvantitet:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30179496 1 g
30179085 500 mg
30179386 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30179496 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM3567301G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 59081-61-7
Molekylformel C10H13NO2
Molekylvikt (g/mol) 179.22
IUPAC-namn 2-anilino-2-methylpropanoic acid
LEDER CC(C)(Nc1ccccc1)C(=O)O
Specifikationer
Kemiskt namn eller material 2-methyl-N-phenylalanine
InChI formel InChI=1S/C10H13NO2/c1-10(2,9(12)13)11-8-6-4-3-5-7-8/h3-7,11H,1-2H3,(H,12,13)
Kvantitet 1 g
Formel vikt 179.0946