missing translation for 'onlineSavingsMsg'
Learn More

2-Methylamino-3-methylpyridine-5-boronic acid pinacol ester, 96%, Thermo Scientific Chemicals

Catalog Number p-7036270
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1111637-92-3
C13H21BN2O2
248.133
MFCD12923387
VZNAKHVRKWXDMU-UHFFFAOYSA-N
n,3-dimethyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-amine, 2-methylamino-3-methylpyridine-5-boronic acid pinacol ester, 2-n-methylamino-3-methylpyridine-5-boronic acid pinacol ester, 5-methyl-6-methylamino pyridine-3-boronic acid pinacol ester, methyl-3-methyl-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridin-2-yl-amine, n,3-dimethyl-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridin-2-amine, n,3-dimethyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-pyridinamine, n-methyl-3-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-amine
57416512
N,3-dimethyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(N=C2)NC)C
This item is not returnable. View return policy
1111637-92-3
C13H21BN2O2
248.133
MFCD12923387
VZNAKHVRKWXDMU-UHFFFAOYSA-N
n,3-dimethyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-amine, 2-methylamino-3-methylpyridine-5-boronic acid pinacol ester, 2-n-methylamino-3-methylpyridine-5-boronic acid pinacol ester, 5-methyl-6-methylamino pyridine-3-boronic acid pinacol ester, methyl-3-methyl-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridin-2-yl-amine, n,3-dimethyl-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridin-2-amine, n,3-dimethyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-pyridinamine, n-methyl-3-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-amine
57416512
N,3-dimethyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(N=C2)NC)C

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Incompatible with oxidizing agents, air, heat. Keep at cool temperature.
TRUSTED_SUSTAINABILITY
Melting Point 105°C to 108°C
Quantity 250 mg
Formula Weight 248.13
Percent Purity 96%
Chemical Name or Material 2-Methylamino-3-methylpyridine-5-boronic acid pinacol ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'