Läs mer
(2-Methylamino-5-nitrophenyl)phenylmethanone, TRC
Produktkod. 30180279
Change view
Kvantitet:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30180279 50 mg
30180081 500 mg
30180152 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30180279 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM21971850MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 4958-56-9
Molekylformel C14H12N2O3
Molekylvikt (g/mol) 256.25
Synonym 2-(Methylamino)-5-nitrobenzophenone
IUPAC-namn [2-(methylamino)-5-nitrophenyl]-phenylmethanone
LEDER CNc1ccc(cc1C(=O)c2ccccc2)[N+](=O)[O-]
Specifikationer
Kemiskt namn eller material (2-Methylamino-5-nitrophenyl)phenylmethanone
Kvantitet 50 mg
InChI formel InChI=1S/C14H12N2O3/c1-15-13-8-7-11(16(18)19)9-12(13)14(17)10-5-3-2-4-6-10/h2-9,15H,1H3
Formel vikt 256.0848