Läs mer
2-(Methylamino)benzamide, TRC
Produktkod. 30479691
Change view
Kvantitet:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30479691 100 mg
30468355 1 g
30464628 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30479691 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM331028100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 7505-81-9
Molekylformel C8H10N2O
Molekylvikt (g/mol) 150.18
Synonym 2-Carbamoyl-N-methylaniline, NSC 401247
IUPAC-namn 2-(methylamino)benzamide
LEDER CNc1ccccc1C(=O)N
Specifikationer
Kemiskt namn eller material 2-(Methylamino)benzamide
Kvantitet 100 mg
InChI formel InChI=1S/C8H10N2O/c1-10-7-5-3-2-4-6(7)8(9)11/h2-5,10H,1H3,(H2,9,11)
Formel vikt 150.0793