Läs mer
2-Methylbenzenesulfonic Acid Dihydrate, TRC
Produktkod. 30475883
Change view
Kvantitet:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30475883 10 g
30474084 1 g
30465916 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30475883 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM28889410G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 68066-37-5
Molekylformel C7H8O3S . 2(H 2O)
Molekylvikt (g/mol) 208.24
Synonym o-Toluenesulfonic Acid Dihydrate
IUPAC-namn 2-methylbenzenesulfonic acid dihydrate
LEDER O=S(O)(C1=C(C)C=CC=C1)=O.O.O
Specifikationer
Kemiskt namn eller material 2-Methylbenzenesulfonic Acid Dihydrate
Kvantitet 10 g
InChI formel InChI=1S/C7H8O3S.2H2O/c1-6-4-2-3-5-7(6)11(8,9)10;;/h2-5H,1H3,(H,8,9,10);2*1H2
Formel vikt 208.04