Läs mer
2-Methylbutanal 2,4-Dinitrophenylhydrazone, TRC
Produktkod. 30472585
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30472585 1 g
30472775 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30472585 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM2662101G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1646-99-7
Molekylformel C11 H14 N4 O4
Molekylvikt (g/mol) 266.25
Synonym 2-Methylbutanal 2-(2,4-Dinitrophenyl)hydrazone, 2-Methylbutyraldehyde (2,4-Dinitrophenyl)hydrazone, α-Methylbutyraldehyde 2,4-Dinitrophenylhydrazone
IUPAC-namn N-(2-methylbutylideneamino)-2,4-dinitro-aniline
LEDER CCC(C)C=NNc1ccc(cc1[N+](=O)[O-])[N+](=O)[O-]
Specifikationer
Kemiskt namn eller material 2-Methylbutanal 2,4-Dinitrophenylhydrazone
Kvantitet 1 g
InChI formel InChI=1S/C11H14N4O4/c1-3-8(2)7-12-13-10-5-4-9(14(16)17)6-11(10)15(18)19/h4-8,13H,3H2,1-2H3
Formel vikt 266.1015