Läs mer
2-Methylcyclohexylmethanol (Mixture of Diastereomers), TRC
Produktkod. 30477328
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30477328 100 mg
1 options
Produktkod. 30477328 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM294985100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-Methylcyclohexylmethanol (Mixture of Diastereomers)
CAS 2105-40-0
Molekylformel C8H16O
Kvantitet 100 mg
Synonym 2-Methyl-cyclohexanemethanol, (2-Methylcyclohexyl)methanol, 2-Methylcyclohexanemethanol, Hexahydro-o-methylbenzyl Alcohol, NSC 19178
InChI formel InChI=1S/C8H16O/c1-7-4-2-3-5-8(7)6-9/h7-9H,2-6H2,1H3
LEDER OCC1C(C)CCCC1
IUPAC-namn (2-methylcyclohexyl)methanol
Molekylvikt (g/mol) 128.21
Formel vikt 128.1201