Läs mer
2-Methyldecane, TRC
Produktkod. 30483143
Change view
Kvantitet:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30483143 100 mg
30481246 1 g
30469834 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30483143 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM091205100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 6975-98-0
Molekylformel C11H24
Molekylvikt (g/mol) 156.31
Synonym Isoundecane, Isopah H, Isopar G
IUPAC-namn 2-methyldecane
LEDER CCCCCCCCC(C)C
Specifikationer
Kemiskt namn eller material 2-Methyldecane
Kvantitet 100 mg
InChI formel InChI=1S/C11H24/c1-4-5-6-7-8-9-10-11(2)3/h11H,4-10H2,1-3H3
Formel vikt 156.1878