Läs mer
2-Methylnaphth[1,2-d]oxazole, TRC
Produktkod. 30468870
Change view
Kvantitet:
500 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30468870 500 mg
30478821 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30468870 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM332348500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-Methylnaphth[1,2-d]oxazole
CAS 85-15-4
Molekylformel C12 H9 N O
Kvantitet 500 mg
Synonym 2-Methyl-4,5-benzobenzoxazole, 2-Methylnaphtho[1,2-d][1,3]oxazole, 2-Methylnaphtho[1,2-d]oxazole
InChI formel InChI=1S/C12H9NO/c1-8-13-12-10-5-3-2-4-9(10)6-7-11(12)14-8/h2-7H,1H3
LEDER Cc1oc2ccc3ccccc3c2n1
IUPAC-namn 2-methylbenzo[e][1,3]benzoxazole
Molekylvikt (g/mol) 183.206
Formel vikt 183.068