Läs mer
(±)-2'-Methylpropranolol Hydrochloride, TRC
Produktkod. 30466500
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30466500 100 mg
1 options
Produktkod. 30466500 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM327190100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (2RS)-1-[(1,1-Dimethylethyl)amino]-3-(naphthalen-1-yloxy)propan-2-ol Hydrochloride
CAS 15230-34-9
Molekylformel C17 H23 N O2 . Cl H
Kvantitet 100 mg
Synonym (2RS)-1-[(1,1-Dimethylethyl)amino]-3-(naphthalen-1-yloxy)propan-2-ol hydrochloride, Nadolol Imp. F (EP) as Hydrochloride, 2-Propanol, 1-[(1,1-dimethylethyl)amino]-3-(1-naphthalenyloxy)-, hydrochloride (1:1), 2-Propanol, 1-(tert-butylamino)-3-(1-naphthyloxy)-, hydrochloride (8CI), 2-Propanol, 1-[(1,1-dimethylethyl)amino]-3-(1-naphthalenyloxy)-, hydrochloride (9CI), 1-tert.-Butylamino-3-(1-naphthoxy)-2-propanol hydrochloride, 1-[(1,1-Dimethylethyl)amino]-3-(1-naphthalenyloxy)-2-propanol hydrochloride, (RS)-2'-Methylpropranolol hydrochloride, (±)-2'-Methylpropranolol hydrochloride
InChI formel InChI=1S/C17H23NO2.ClH/c1-17(2,3)18-11-14(19)12-20-16-10-6-8-13-7-4-5-9-15(13)16;/h4-10,14,18-19H,11-12H2,1-3H3;1H
LEDER Cl.CC(C)(C)NCC(O)COc1cccc2ccccc12
IUPAC-namn 1-(tert-butylamino)-3-naphthalen-1-yloxypropan-2-ol;hydrochloride
Molekylvikt (g/mol) 309.83
Formel vikt 309.1496