Läs mer
2-methylquinoline-4,6-diamine, TRC
Produktkod. 30475229
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30475229 50 mg
30465177 100 mg
30479434 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30475229 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM35650550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 5443-31-2
Molekylformel C10H11N3
Molekylvikt (g/mol) 173.22
IUPAC-namn 2-methylquinoline-4,6-diamine
LEDER Cc1cc(N)c2cc(N)ccc2n1
Specifikationer
Kemiskt namn eller material 2-methylquinoline-4,6-diamine
Kvantitet 50 mg
InChI formel InChI=1S/C10H11N3/c1-6-4-9(12)8-5-7(11)2-3-10(8)13-6/h2-5H,11H2,1H3,(H2,12,13)
Formel vikt 173.0953