Läs mer
2-Methylsulfinylethyl-2-methylthioethylsulfone-13C4, TRC
Produktkod. 30478685
Change view
Kvantitet:
2.5 mg
5 mg
Förpackningsstorlek:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30478685 2.5 mg 2.5mg
30478684 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30478685 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM2648572.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-Methylsulfinylethyl-2-methylthioethylsulfone-13C4
CAS 672310-54-2
Molekylformel C213C4H14O3S3
Kvantitet 2.5 mg
Synonym 1-[[2-(Methylsulfinyl)ethyl-13C2]sulfonyl]-2-(methylthio)-ethane-13C2
InChI formel InChI=1S/C6H14O3S3/c1-10-3-5-12(8,9)6-4-11(2)7/h3-6H2,1-2H3/i3+1,4+1,5+1,6+1
LEDER CS[13CH2][13CH2]S([13CH2][13CH2]S(C)=O)(=O)=O
Molekylvikt (g/mol) 234.34