missing translation for 'onlineSavingsMsg'
Läs mer

2-Methylsulfonyl-4-trifluoromethylbenzoic Acid, TRC

Produktkod. 30478494
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30478494 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30478494 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM32813525MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material 2-Methylsulfonyl-4-trifluoromethylbenzoic Acid
CAS 142994-06-7
Molekylformel C9 H7 F3 O4 S
Kvantitet 25 mg
Synonym Benzoic acid, 2-(methylsulfonyl)-4-(trifluoromethyl)-, 2-Methylsulfonyl-4-trifluoromethylbenzoic acid, AE B197555, RPA 203328
InChI formel InChI=1S/C9H7F3O4S/c1-17(15,16)7-4-5(9(10,11)12)2-3-6(7)8(13)14/h2-4H,1H3,(H,13,14)
LEDER CS(=O)(=O)c1cc(ccc1C(=O)O)C(F)(F)F
IUPAC-namn 2-methylsulfonyl-4-(trifluoromethyl)benzoic acid
Molekylvikt (g/mol) 268.21
Formel vikt 268.0017
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.