missing translation for 'onlineSavingsMsg'
Learn More

2-Methylsulfonylpyrimidine-5-carboxylic Acid, TRC

Product Code. 30470551
Change view
Click to view available options
Kvantitet:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30470551 1 g
30466048 500 mg
30471737 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30470551 Supplier Toronto Research Chemicals Supplier No. TRCM2029011G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 914208-17-6
Molekylformel C6 H6 N2 O4 S
Molekylvikt (g/mol) 202.188
Synonym 2-(Methylsulfonyl)-5-pyrimidinecarboxylic Acid, 2-Methanesulfonyl-pyrimidine-5-carboxylic Acid, 2-Methanesulfonylpyrimidine-5-carboxylic Acid
IUPAC-namn 2-methylsulfonylpyrimidine-5-carboxylic acid
LEDER CS(=O)(=O)c1ncc(cn1)C(=O)O

Specifications

Kemiskt namn eller material 2-Methylsulfonylpyrimidine-5-carboxylic Acid
Kvantitet 1 g
InChI formel InChI=1S/C6H6N2O4S/c1-13(11,12)6-7-2-4(3-8-6)5(9)10/h2-3H,1H3,(H,9,10)
Formel vikt 202.005
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.