Läs mer
2-Methylsulfonylpyrimidine-5-carboxylic Acid, TRC
Produktkod. 30471737
Change view
Kvantitet:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30471737 100 mg
30470551 1 g
30466048 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30471737 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM202901100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 914208-17-6
Molekylformel C6 H6 N2 O4 S
Molekylvikt (g/mol) 202.188
Synonym 2-(Methylsulfonyl)-5-pyrimidinecarboxylic Acid, 2-Methanesulfonyl-pyrimidine-5-carboxylic Acid, 2-Methanesulfonylpyrimidine-5-carboxylic Acid
IUPAC-namn 2-methylsulfonylpyrimidine-5-carboxylic acid
LEDER CS(=O)(=O)c1ncc(cn1)C(=O)O
Specifikationer
Kemiskt namn eller material 2-Methylsulfonylpyrimidine-5-carboxylic Acid
Kvantitet 100 mg
InChI formel InChI=1S/C6H6N2O4S/c1-13(11,12)6-7-2-4(3-8-6)5(9)10/h2-3H,1H3,(H,9,10)
Formel vikt 202.005