Läs mer
2-Methylthiazolidine-4-carboxylic Acid, TRC
Produktkod. 30464838
Change view
Kvantitet:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30464838 100 mg
30479163 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30464838 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM330300100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 4165-32-6
Molekylformel C5H9NO2S
Molekylvikt (g/mol) 147.2
Synonym 2-Methyl-4-thiazolidinecarboxylic Acid
IUPAC-namn 2-methyl-1,3-thiazolidine-4-carboxylic acid
LEDER CC1NC(CS1)C(=O)O
Specifikationer
Kemiskt namn eller material 2-Methylthiazolidine-4-carboxylic Acid
Kvantitet 100 mg
InChI formel InChI=1S/C5H9NO2S/c1-3-6-4(2-9-3)5(7)8/h3-4,6H,2H2,1H3,(H,7,8)
Formel vikt 147.0354