missing translation for 'onlineSavingsMsg'
Läs mer

2-(Methylthio)benzothiazole, TRC

Produktkod. 30480294
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
250 mg
500 mg
2.5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30480294 500 mg
30470514 2.5 g
30480295 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30480294 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM725628500MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 615-22-5
Molekylformel C8 H7 N S2
Molekylvikt (g/mol) 181.28
Synonym Benzothiazole, 2-(methylthio)- (6CI, 7CI, 8CI, 9CI, ACI), 2-(Methylthio)benzothiazole (ACI), 2-(Methylmercapto)benzothiazole, 2-(Methylthio)benzo[d]thiazole, 2-Methylsulfanyl-1,3-benzothiazole, Methylcaptax, NSC 31259, NSC 41046, NSC 5352
IUPAC-namn 2-methylsulfanyl-1,3-benzothiazole
LEDER CSc1nc2ccccc2s1

Specifications

Kemiskt namn eller material 2-(Methylthio)benzothiazole
Kvantitet 500 mg
InChI formel InChI=1S/C8H7NS2/c1-10-8-9-6-4-2-3-5-7(6)11-8/h2-5H,1H3
Formel vikt 181.002
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.