missing translation for 'onlineSavingsMsg'
Learn More

2-(Methylthio)benzylamine, 95%, Thermo Scientific™

Catalog Number p-2696439
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
56004-83-2
C8H11NS
153.243
MFCD00831039
YVEJLBIEESKJLG-UHFFFAOYSA-N
2-methylthio benzylamine, 2-methylsulfanyl phenyl methanamine, 2-methylthio phenyl methanamine, 1-2-methylsulfanyl phenyl methanamine, 2-methylsulfanylphenyl methanamine, 2-methylthiobenzylamine, 2-methylthio benzyl amine, 2-methylthio-benzyl amine, 2-methylthiophenyl methylamine
2736981
(2-methylsulfanylphenyl)methanamine
CSC1=CC=CC=C1CN
This item is not returnable. View return policy
56004-83-2
C8H11NS
153.243
MFCD00831039
YVEJLBIEESKJLG-UHFFFAOYSA-N
2-methylthio benzylamine, 2-methylsulfanyl phenyl methanamine, 2-methylthio phenyl methanamine, 1-2-methylsulfanyl phenyl methanamine, 2-methylsulfanylphenyl methanamine, 2-methylthiobenzylamine, 2-methylthio benzyl amine, 2-methylthio-benzyl amine, 2-methylthiophenyl methylamine
2736981
(2-methylsulfanylphenyl)methanamine
CSC1=CC=CC=C1CN
CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber glass bottle
Linear Formula CH3SC6H4CH2NH2
Quantity 250 mg
Formula Weight 153.25
Percent Purity 95%
Chemical Name or Material 2-(Methylthio)benzylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'