Läs mer
2-Methylvaleraldehyde (>90%), TRC
Produktkod. 30477067
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30477067 1 mg
1 options
Produktkod. 30477067 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM3397681MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-Methylvaleraldehyde
CAS 123-15-9
Molekylformel C6H12O
Kvantitet 1 mg
Synonym 2-Methylpentanal (ACI), Valeraldehyde, 2-methyl- (6CI, 7CI, 8CI), (±)-2-Methylpentanal, 2-Formylpentane, 2-Methylvaleraldehyde, 2-Methylvaleric aldehyde, NSC 49351, α-Methylpentanal, α-Methylvaleraldehyde
InChI formel InChI=1S/C6H12O/c1-3-4-6(2)5-7/h5-6H,3-4H2,1-2H3
LEDER CCCC(C)C=O
IUPAC-namn 2-methylpentanal
Molekylvikt (g/mol) 100.16
Formel vikt 100.0888