missing translation for 'onlineSavingsMsg'
Learn More

2-n-Butyl-5-ethylthiophene, 97%, Thermo Scientific™

Catalog Number p-7026618
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
54411-06-2
C10H16S
168.298
MFCD00053118
ORAPMLHUSQJCQH-UHFFFAOYSA-N
2-n-butyl-5-ethylthiophene, thiophene, 2-butyl-5-ethyl, 2-butyl-5-ethyl-thiophene, #
521504
2-butyl-5-ethylthiophene
CCCCC1=CC=C(S1)CC
This item is not returnable. View return policy
54411-06-2
C10H16S
168.298
MFCD00053118
ORAPMLHUSQJCQH-UHFFFAOYSA-N
2-n-butyl-5-ethylthiophene, thiophene, 2-butyl-5-ethyl, 2-butyl-5-ethyl-thiophene, #
521504
2-butyl-5-ethylthiophene
CCCCC1=CC=C(S1)CC

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 5 g
Formula Weight 168.3
Percent Purity 97%
Chemical Name or Material 2-n-Butyl-5-ethylthiophene
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'