Learn More
2-n-Hexyldecanoic acid, 97%
Product Code. 11318695
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11318695 100 g 100g
11308695 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11318695 Supplier Thermo Scientific Alfa Aesar Supplier No. L12238.22
 Restricted Item
I lager
Add to basket
CAS: 25354-97-6 | C16H32O2 | 256.43 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 25354-97-6
Molecular Formula C16H32O2
Molecular Weight (g/mol) 256.43
MDL Number MFCD00059651
InChI Key JMOLZNNXZPAGBH-UHFFFAOYSA-N
Synonym decanoic acid, 2-hexyl, hexyldecanoic acid, c16 guerbet fatty acid, 2-hexyldecansaeure, 7-pentadecanecarboxylic acid, 2-hexyldecansaeure german, 2-hexyl-decanoic acid, 2-n-hexyldecanoic acid, 2-hexyldecanoicacid, isocarb 16
PubChem CID 32912
IUPAC Name 2-hexyldecanoic acid
SMILES CCCCCCCCC(CCCCCC)C(=O)O
Specifications
Melting Point 10°C
Density 0.88
Boiling Point 165°C to 168°C (2 mmHg)
Flash Point 174°C (345°F)
Refractive Index 1.446
Quantity 100 g
Beilstein 1777217
Merck Index 14,4709
Formula Weight 256.43
Percent Purity 97%
Chemical Name or Material 2-n-Hexyldecanoic acid