missing translation for 'onlineSavingsMsg'
Learn More

2-n-Propyl-1-pentanol, 97%, Thermo Scientific Chemicals

Catalog Number p-7052887
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
58175-57-8
C8H18O
130.231
MFCD00010260
LASHFHLFDRTERB-UHFFFAOYSA-N
2-propyl-1-pentanol, 1-pentanol, 2-propyl, 2-propylpentanol, 2-n-propyl-1-pentanol, acmc-1azt3, 2-propyl-1-hydroxypentane
123543
2-propylpentan-1-ol
CCCC(CCC)CO
This item is not returnable. View return policy
58175-57-8
C8H18O
130.231
MFCD00010260
LASHFHLFDRTERB-UHFFFAOYSA-N
2-propyl-1-pentanol, 1-pentanol, 2-propyl, 2-propylpentanol, 2-n-propyl-1-pentanol, acmc-1azt3, 2-propyl-1-hydroxypentane
123543
2-propylpentan-1-ol
CCCC(CCC)CO

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 0.83 g/mL
Boiling Point 80°C to 82°C (12 mmHg)
Flash Point 72°C (162°F)
Linear Formula (CH3CH2CH2)2CHCH2OH
Refractive Index 1.43
Quantity 1 g
Formula Weight 130.23
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-n-Propyl-1-pentanol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'