Läs mer
2-n-Propyl-4-methyl-6-(1-methylbenzimidazol-2-yl)-benzimidazole, TRC
Produktkod. 30003494
Change view
Kvantitet:
50 mg
100 mg
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30003494 5 g
17953595 100 mg
17912905 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30003494 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP8382805G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 152628-02-9
Molekylformel C19 H20 N4
Molekylvikt (g/mol) 304.39
Synonym 4-Methyl-6-(1-methyl-1H-benzimidazol-2-yl)-2-propyl-1H-benzimidazole, Telmisartan Imp. A (EP), Telmisartan Related Compound A (USP)
IUPAC-namn 4-methyl-6-(1-methylbenzimidazol-2-yl)-2-propyl-1H-benzimidazole
LEDER CCCc1nc2c(C)cc(cc2[nH]1)c3nc4ccccc4n3C
Specifikationer
Kemiskt namn eller material 4-Methyl-6-(1-methyl-1H-benzimidazol-2-yl)-2-propyl-1H-benzimidazole
Anteckningar om renhetsgrad HPLC
Kvantitet 5 g
InChI formel InChI=1S/C19H20N4/c1-4-7-17-20-15-11-13(10-12(2)18(15)22-17)19-21-14-8-5-6-9-16(14)23(19)3/h5-6,8-11H,4,7H2,1-3H3,(H,20,22)
Formel vikt 304.1688
Procent renhet >95%