missing translation for 'onlineSavingsMsg'
Learn More

2-n-Propylpyridine, 97%, Thermo Scientific™

Catalog Number p-7077352
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
622-39-9
C8H11N
121.18
OIALIKXMLIAOSN-UHFFFAOYSA-N
2-n-propylpyridine, conyrine, conyrin, pyridine, 2-propyl, 1-2-pyridyl propane, 2-propyl-pyridine, unii-3n6ax0b7pa, 2-propyl pyridine, pyridine, propyl, pyridine, 1-propyl
69320
2-propylpyridine
CCCC1=CC=CC=N1
This item is not returnable. View return policy
622-39-9
C8H11N
121.18
OIALIKXMLIAOSN-UHFFFAOYSA-N
2-n-propylpyridine, conyrine, conyrin, pyridine, 2-propyl, 1-2-pyridyl propane, 2-propyl-pyridine, unii-3n6ax0b7pa, 2-propyl pyridine, pyridine, propyl, pyridine, 1-propyl
69320
2-propylpyridine
CCCC1=CC=CC=N1
Density 0.9120g/mL
Boiling Point 169.0°C to 171.0°C
Flash Point 56°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 5 g
Specific Gravity 0.912
Formula Weight 121.18
Percent Purity 97%
Chemical Name or Material 2-n-Propylpyridine
compliance-icons compliance-icons
Product Identifier
  • 2-n-Propylpyridine, 97%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'