missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-n-Propylthiazolidine-4-carboxylic acid, 97%

Brand:  Alfa Aesar™ H62991.06

Additional Details : CAS Number : 4165-34-8 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15479418

  • 9429.38 SEK / 5g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

2-n-Propylthiazolidine-4-carboxylic acid
4165-34-8
175.25
MFCD09930114

Specifications

2-n-Propylthiazolidine-4-carboxylic acid
4165-34-8
97%
MFCD09930114
5g
Solid
175.25
Videos
SDS